Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADP-Ribosylation Factor 6 Mouse anti-Human, Clone: 6ARF01 , MilliporeSigma™

Catalog No. 051149MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-114-9MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-114-9MI Supplier MilliporeSigma Supplier No. 051149

Mouse Monoclonal Antibody

Anti-ADP-Ribosylation Factor 6 Antibody, clone 6ARF01 is a Mouse Monoclonal Antibody for detection of ADP-Ribosylation Factor 6 also known as ADP-ribosylation factor 6 and has been validated in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADP-Ribosylation Factor 6
Applications Western Blot
Classification Monoclonal
Clone 6ARF01
Conjugate Unconjugated
Formulation Purified monoclonal IgG ascites in 10mM PBS, pH 7.4, with 0.2% BSA and 0.09% sodium azide.
Gene Accession No. P62330
Gene Symbols DKFZp564M0264
Host Species Mouse
Immunogen Recombinant human ARF6 protein.
Purification Method Protein G chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.