Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADPRH, Mouse, Clone: 1F11, Abnova™

Catalog No. 89016058 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-058 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-058 Supplier Abnova Corporation Supplier No. H00000141M04
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant ADPRH.

The enzyme encoded by this gene catalyzes removal of mono-ADP-ribose from arginine residues of proteins in the ADP-ribosylation cycle. Unlike the rat and mouse enzymes, which require DTT for maximal activity, the human enzyme is DTT-independent. [provided by RefSeq

Sequence: KWEFLQDGEKIHRQLAQLGGLDALDVGRWRVSDDTVMHLATAEALVEAGKAPKLTQLYYLLAKHYQDCMEDMDGRAPGGASVHNAMQLKPGKPNGWRIP

Specifications

Antigen ADPRH
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 1F11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant ADPRH.
Formulation PBS with no preservative; pH 7.4
Gene ADPRH
Gene Accession No. NM_001125
Gene Alias ARH1
Gene Symbols ADPRH
Host Species Mouse
Immunogen ADPRH (NP_001116.1, 23 a.a. ∼ 121 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 141
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.