Learn More
ADRA1A, Mouse, Clone: 4C7, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ADRA1A.
Supplier: Abnova Corporation H00000148M01
Description
Alpha-1-adrenergic receptors (alpha-1-ARs) are members of the G protein-coupled receptor superfamily. They activate mitogenic responses and regulate growth and proliferation of many cells. There are 3 alpha-1-AR subtypes: alpha-1A, -1B and -1D, all of which signal through the Gq/11 family of G-proteins and different subtypes show different patterns of activation. This gene encodes alpha-1A-adrenergic receptor. Alternative splicing of this gene generates four transcript variants, which encode four different isoforms with distinct C-termini but having similar ligand binding properties. [provided by RefSeq
Sequence: MVFLSGNASDSSNCTQPPAPVNISKAISpecifications
ADRA1A | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_033303 | |
ADRA1A | |
ADRA1A (NP_150646, 1 a.a. ∼ 27 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Western Blot | |
4C7 | |
Mouse monoclonal antibody raised against a partial recombinant ADRA1A. | |
ADRA1A | |
ADRA1C/ADRA1L1/ALPHA1AAR | |
Mouse | |
Affinity chromatography | |
RUO | |
148 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.