Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADRA1A, Mouse, Clone: 5G8, Abnova™

Catalog No. 89016060 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-060 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-060 Supplier Abnova Corporation Supplier No. H00000148M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant ADRA1A.

Alpha-1-adrenergic receptors (alpha-1-ARs) are members of the G protein-coupled receptor superfamily. They activate mitogenic responses and regulate growth and proliferation of many cells. There are 3 alpha-1-AR subtypes: alpha-1A, -1B and -1D, all of which signal through the Gq/11 family of G-proteins and different subtypes show different patterns of activation. This gene encodes alpha-1A-adrenergic receptor. Alternative splicing of this gene generates four transcript variants, which encode four different isoforms with distinct C-termini but having similar ligand binding properties. [provided by RefSeq

Sequence: MVFLSGNASDSSNCTQPPAPVNISKAI

Specifications

Antigen ADRA1A
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5G8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant ADRA1A.
Formulation PBS with no preservative; pH 7.4
Gene ADRA1A
Gene Accession No. NM_033303
Gene Alias ADRA1C/ADRA1L1/ALPHA1AAR
Gene Symbols ADRA1A
Host Species Mouse
Immunogen ADRA1A (NP_150646, 1 a.a. ∼ 27 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.