Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adrenocorticotropic Hormone Mouse anti-Human, Rat, Clone: 57, MilliporeSigma™

Catalog No. CBL57MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL57MI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL57MI Supplier MilliporeSigma Supplier No. CBL57
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Adrenocorticotropic Hormone Antibody, N-terminus, clone 57 is validated for use in ELISA, immunohistochemistry (paraffin) for the detection of Adrenocorticotropic Hormone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adrenocorticotropic Hormone
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 57
Conjugate Unconjugated
Formulation The monoclonal is presented as 1mg in 1ml phosphate buffered saline containing 10mM sodium azide. We recommend that each laboratory determine an optimum working titre for use in its particular application.
Gene Accession No. NM_000939.2; NM_001035256.1
Gene Symbols POMC, pro-ACTH-endorphin, adrenocorticotropin, beta-LPH, POC, gamma-LPH, LPH, ACTH, corticotropin, beta-endorphin, beta-MSH, alpha-MSH, pro-opiomelanocortin, proopiomelanocortin, Pro-opiomelanocortin, corticotropin-lipotrophin, NPP, MSH, met-enkephalin, C
Host Species Mouse
Quantity 1 mg
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage 4°C for up to 1 month, −20°C in undiluted aliquots for long term
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.