Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adrenoleukodystrophy Protein Mouse anti-Human, Clone: 2AL-1D6, MilliporeSigma™

Catalog No. MAB2164MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2164MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2164MI Supplier MilliporeSigma Supplier No. MAB2164
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Adrenoleukodystrophy Protein using this Anti-Adrenoleukodystrophy Protein Antibody, a.a. 279-482, clone 2AL-1D6 validated for use in ELISA, western blot, immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adrenoleukodystrophy Protein
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2AL-1D6
Conjugate Unconjugated
Formulation Ascites. Liquid, does not contain any preservative.
Gene Accession No. NM_000033.2
Gene Symbols ABCD1, ALD, AMN, adrenoleukodystrophy/adrenomyeloneuropathy, ALDP, X-ALD, ABC42, adrenoleukodystrophy
Host Species Mouse
Immunogen ALDP1 fragment from aa 279 to 482 as a fusion protein
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.