Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-AFP Clone: 1G7,

Catalog No. ST1673100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ST1673100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ST1673100UG Supplier MilliporeSigma™ Supplier No. ST1673100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects AFP Clone: 1G7 in Human samples, and it is validated for ELISA, Immunocytochemistry, Immuno Blotting

This gene encodes α-fetoprotein, a major plasma protein produced by the yolk sac and the liver during fetal life. α-fetoprotein expression in adults is often associated with hepatoma or teratoma. However, hereditary persistence of α-fetoprotein may also be found in individuals with no obvious pathology. The protein is thought to be the fetal counterpart of serum albumin, and the α-fetoprotein and albumin genes are present in tandem in the same transcriptional orientation on chromosome 4. α-fetoprotein is found in monomeric as well as dimeric and trimeric forms, and binds copper, nickel, fatty acids and bilirubin. The level of α-fetoprotein in amniotic fluid is used to measure renal loss of protein to screen for spina bifida and anencephaly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AFP
Applications ELISA, Immunoblot, Immunoblot, Immunocytochemistry
Classification Monoclonal
Clone 1G7
Conjugate Unconjugated
Formulation In PBS, pH 7.2.
Host Species Mouse
Immunogen A recombinant polypeptide corresponding to amino acids of 500-610 human AFP, expressed as a GST fusion protein
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽70-75kDa AFP protein in HepG2 cells.
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.