Learn More
Description
This gene encodes a member of the immunoglobulin superfamily of cell surface molecules. It is a receptor for various molecules, including the amyloidogenic form of serum amyloid A, amyloid-beta protein, members of the S100/calgranulin superfamily and advanced glycation end products. The gene lies within the major histocompatibility complex (MHC) class III region on chromosome 6. Alternative splicing results in two transcript variants encoding different isoforms. [provided by RefSeq
Specifications
Specifications
| Antigen | AGER |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1C1 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a full length recombinant AGER. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | AGER |
| Gene Accession No. | BC020669 |
| Gene Alias | MGC22357/RAGE |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.