Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aggrecan Rabbit anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB1031MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1031MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1031MI Supplier MilliporeSigma Supplier No. AB1031
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Aggrecan Antibody is an antibody against Aggrecan for use in immunofluoresence, immunohistochemistry (paraffin), immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aggrecan
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal antigen affinity purified antibody in PBS. Anti-GST has not been adsorbed. Liquid.
Note: Aggrecan is present in articular cartilage, brain, heart, and other tissues.
Gene Symbols ACAN; MSK16; SEDK; CSPCP; AGCAN; AGC1; CSPG1; CSPGCP
Host Species Rabbit
Immunogen GST fusion protein containing amino acids 1177-1326 of mouse aggrecan.
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_001126
Target Species Mouse
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.