Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aggregated a-Synuclein Mouse anti-Human, Clone: 5G4, MilliporeSigma™

Catalog No. MABN389MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN389MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN389MI Supplier MilliporeSigma Supplier No. MABN389
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Aggregated a-Synuclein Antibody, clone 5G4 is validated for use in western blotting, immunohistochemistry and ELISA for the detection of Aggregated a-Synuclein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aggregated a-Synuclein
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 5G4
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing PBS without preservatives.
Gene Accession No. NP_000336
Gene Symbols SNCA, NACP, PARK1
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to human Aggregated a-Synuclein.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.