Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Agmatine Rabbit anti-Vertebrates, Polyclonal

Catalog No. AB15682000T Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB15682000T 1 Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB15682000T Supplier MilliporeSigma™ Supplier No. AB15682000T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Agmatine Antibody detects level of Agmatine and has been published and validated for use in immunocytochemistry, immunohistochemistry (paraffin) and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Agmatine Antibody
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit antisera. Liquid in 100mM phosphate buffer with 1% goat serum and 0.05% thimerosal.
Host Species Rabbit
Immunogen Agmatine conjugated to BSA
Quantity 1 Unit
Primary or Secondary Primary
Target Species Vertebrates
Content And Storage 2-8°C in undiluted aliquotes for up to 6 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.