Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ago family Mouse anti-Human, MilliporeSigma™

Catalog No. 04085MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-085-MI 1 Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-085-MI Supplier MilliporeSigma Supplier No. 04085
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-Ago family Antibody (Mouse Monoclonal Antibody) validated in western blotting to detect Ago family also known as argonaute family PIWI.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ago family
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation 100μg of Protein A purified mouse monoclonal IgG1κ in 100μL of 1X PBS, pH 7.0, 0.1% Azide.
Gene Accession No. NM_012154.2; NM_012199.2
Gene Symbols EIF2C1, AGO1, EIF2C, Argonaute-1, GERP95, Q99, DKFZp686M13167
Host Species Mouse
Immunogen Synthetic peptide PKIDVYHYEVDIKPDKCPRRVNREVV
Purification Method Protein A purfied
Quantity 1 Unit
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.