Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Ago1 Clone: 4B8,

Catalog No. MABE1121MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABE1121MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE1121MI Supplier MilliporeSigma™ Supplier No. MABE1121
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specifically detects Ago1 Clone: 4B8 in Human samples, and it is validated for Western Blotting

Protein argonaute-1 (UniProt Q9UL18; also known as Argonaute1, Argonaute RISC catalytic component 1, eIF-2C 1, eIF2C 1, Eukaryotic translation initiation factor 2C 1, hAgo1, Putative RNA-binding protein Q99) is encoded by the AGO1 (also known as EIF2C, EIF2C1) gene (Gene ID 26523) in human. Ago1 belongs to the family of argonaute proteins involved in gene expression by regulating gene transcription and RNA splicing in the nucleus, as well as by regulating posttranscriptional gene silencing mediated by siRNAs and microRNAs in the cytosol. Ago1 is shown to interact with chromatin modifiers, splicing factors, and RNA polymerase II. Ago1 and the related Ago2 are also shown to target Hepatitis C Virus (HCV) RNÁ 5'-untranslated region (5'-UTR) in an miR-122-dependent manner, resulting in viral RNA stability and translation stimulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ago1
Applications Western Blot
Classification Monoclonal
Clone 4B8
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q9UL18
Gene Symbols AGO1, EIF2C, EIF2C1
Host Species Rat
Immunogen GST-tagged recombinant human Ago1.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics, Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_036331.1
Test Specificity Clone 4B8 sepcifically immunoprecipitated FLAG-tagged Ago1,but not FLAG-tagged Ago2, Ago3, or Ago4 (Ender, C., et al. (2008). Mol. Cell. 32(4):519-528).
Target Species Human
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.