Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ago2 Rat anti-Human, Clone: 11A9, MilliporeSigma™

Catalog No. MABE253MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE253MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE253MI Supplier MilliporeSigma Supplier No. MABE253
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Use Anti-Ago2 Antibody, clone 11A9 (Rat Monoclonal Antibody) validated in western blot, immunoprecipitation, immunocytochemistry to detect Ago2 also known as Protein argonaute-2, Argonaute2, hAgo2, eIF-2C 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ago2
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 11A9
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001158095
Gene Symbols EIF2C2, AGO2
Host Species Rat
Immunogen Linear peptide corresponding to human Ago2.
Purification Method Protein G purified
Quantity 200 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.