Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Ago4 Mouse anti-Human, Clone: 1B7-G11, MilliporeSigma™

Catalog No. MABE139MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE139MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE139MI Supplier MilliporeSigma Supplier No. MABE139

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Anti-human Ago4 Antibody, clone 1B7-G11 is a Mouse Monoclonal Antibody for detection of human Ago4 clone 1B7-G11 also known as Protein argonaute-4, Argonaute4, hAgo4 and has been validated in western blotting and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ago4
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1B7-G11
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_060099
Gene Symbols EIF2C4, AGO4, KIAA1567
Host Species Mouse
Immunogen Recombinant protein corresponding to human Ago4.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.