Learn More
1-acylglycerol-3-phosphate O-acyltransferase 3, Mouse, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
The protein encoded by this gene is an acyltransferase that converts lysophosphatidic acid into phosphatidic acid, which is the second step in the de novo phospholipid biosynthetic pathway. The encoded protein may be an integral membrane protein. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq
Specifications
Specifications
| Antigen | 1-acylglycerol-3-phosphate O-acyltransferase 3 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a partial recombinant AGPAT3. |
| Formulation | 50% glycerol |
| Gene | AGPAT3 |
| Gene Accession No. | NM_020132 |
| Gene Alias | LPAAT-GAMMA1/MGC4604 |
| Gene Symbols | AGPAT3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.