Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agrin Mouse anti-Mouse, Rat, MilliporeSigma™

Catalog No. MAB5204MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5204MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5204MI Supplier MilliporeSigma Supplier No. MAB5204
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Agrin Antibody is an antibody against Agrin for use in western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Agrin
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in PBS containing 50% glycerol and 0.09% sodium azide.
Gene Accession No. NM_198576.2
Gene Symbols AGRN, agrin, OTTHUMP00000044043, FLJ45064, AGRIN
Host Species Mouse
Immunogen Recombinant rat agrin (C-terminal construct)
Quantity 200 μg
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.