Learn More
AHR, Mouse, Clone: 3B7, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant AHR.
Supplier: Abnova Corporation H00000196M09
Description
This gene encodes a ligand-activated transcription factor involved in the regulation of biological responses to planar aromatic hydrocarbons. This receptor has been shown to regulate xenobiotic-metabolizing enzymes such as cytochrome P450. Its ligands included a variety of aromatic hydrocarbons. [provided by RefSeq
Sequence: EIRTKNFIFRTKHKLDFTPIGCDAKGRIVLGYTEAELCTRGSGYQFIHAADMLYCAESHIRMIKTGESGMIVFRLLTKNNRWTWVQSNARLLYKNGRPSpecifications
| AHR | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001621 | |
| AHR | |
| AHR (NP_001612, 279 a.a. ∼ 376 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA | |
| 3B7 | |
| Mouse monoclonal antibody raised against a full length recombinant AHR. | |
| AHR | |
| bHLHe76 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 196 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.