Learn More
Description
This gene encodes a ligand-activated transcription factor involved in the regulation of biological responses to planar aromatic hydrocarbons. This receptor has been shown to regulate xenobiotic-metabolizing enzymes such as cytochrome P450. Its ligands included a variety of aromatic hydrocarbons. [provided by RefSeq
Specifications
Specifications
| Antigen | aryl hydrocarbon receptor |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 3H4 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant AHR. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | AHR |
| Gene Accession No. | NM_001621 |
| Gene Alias | bHLHe76 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.