Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIM-1 Mouse, Unlabeled, Clone: 6, BD

Catalog No. BDB611083 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611083 150 μg
BDB611082 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611083 Supplier BD Biosciences Supplier No. 611083
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The mitotic phase of the cell cycle is a complex process that ensures the fidelity of chromosome segregation. During the final stage of mitosis(telophase), segregated chromosomes become less condense and nuclear membranes surround the two sets of daughter chromosomes. Simultaneously, the separation and segregation of the cytoplasm (cytokinesis) ensures complete division and formation of two identical daughter cells. Regulation of cytokinesis is poorly understood and errors in this process can lead to cell death or oncogenesis. The Drosophila serine/threonine protein kinase Aurora and the S. cerevisiae Ipl1 kinase are highly homologous and are required for progression through mitosis. Their mammalian homolog AIM-1 (also known as Aurora and Ipl1-like midbody associated protein) accumulates at the G2/M interface. During late anaphase, AIM-1 is found at the equator of central spindles. However, during telophase and cytokinesis, it is found at the midbody. Although over-expression of a kinase-inactive AIM-1 mutant disrupts formation of the cleavage furrow, nuclear division is unaffected. Thus, it is thought that AIM-1 is essential for cleavage furrowing and the onset of cytokinesis.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen AIM-1
Applications Western Blot
Classification Monoclonal
Clone 6
Concentration 250μg/mL
Conjugate Unconjugated
Description Aurora B; Aurora and Ipl1-like midbody associated protein
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat AIM-1 aa. 2-124
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.