Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aiolos Mouse anti-Human, Alexa Fluor 488, Clone: S50-895, BD

Catalog No. BDB565216 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565216 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565216 Supplier BD Biosciences Supplier No. 565216
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The S50-895 monoclonal antibody specifically recognizes the transcription factor, Aiolos, which is encoded by IKZF3 , IKAROS family zinc finger 3. Aiolos belongs to the Ikaros family of zinc-finger proteins that includes Ikaros and Helios. Aiolos can form homodimers or heterodimers with other Ikaros family members. Multiple Aiolos isoforms have been described. Aiolos is expressed in committed lymphoid progenitors and mature types of T and B lymphocytes, but is not expressed by hematopoietic stem cells. Aiolos plays an important role in the control of B lymphocyte differentiation and proliferation and is specifically required for the generation of long-lived, high affinity plasma cells in the bone marrow. Aiolos is involved in regulating BCL2 expression and controlling apoptosis in T-cells in an IL2-dependent manner. Aberrant expression of IKZF3 has been implicated with leukemogenesis and certain autoimmune diseases.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Aiolos
Applications Flow Cytometry
Classification Monoclonal
Clone S50-895
Conjugate Alexa Fluor 488
Description AIOLOS; AIO; IKZF3; ZNFN1A3
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Host Species Mouse
Immunogen Human Aiolos Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.