Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIP1/WD repeat-containing protein 1 Rat anti-Mouse, Clone: 13F10, MilliporeSigma™

Catalog No. MABF264MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF264MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF264MI Supplier MilliporeSigma Supplier No. MABF264
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This Anti-AIP1/WD repeat-containing protein 1, clone 13F10 Antibody is validated for use in Western Blotting for the detection of AIP1/WD repeat-containing protein 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AIP1/WD repeat-containing protein 1
Applications Western Blot
Classification Monoclonal
Clone 13F10
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_035845
Gene Symbols Wdr1
Host Species Rat
Immunogen Recombinant protein corresponding to mouse AIP1/WD repeat-containing protein 1.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.