Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIRE Rabbit anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. 09456MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-456-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-456-MI Supplier MilliporeSigma Supplier No. 09456
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect AIRE using this Anti-AIRE Antibody validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AIRE
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 100 μg of immunoaffinity purified rabbit polyclonal IgG in 173μL of 0.02M PBS, 0.25M NaCl containing 0.1% sodium azide
Gene Symbols AIRE; AIRE1; PGA1; APSI; APS1; PGAI; APECED
Host Species Rabbit
Immunogen KLH-conjugated synthetic peptide corresponding to amino acids 162-181 of mouse AIRE
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_000383.2; NM_000658.1
Target Species Mouse
Content And Storage −20°C for 24 months. Aliquot to avoid repeated freezing and thawing
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.