Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenylate Kinase 3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA515426
Description
AK3 is a member of the adenylate kinase family of enzymes. This protein is localized to the mitochondrial matrix. Adenylate kinases regulate the adenine and guanine nucleotide compositions within a cell by catalyzing the reversible transfer of phosphate group among these nucleotides. Five isozymes of adenylate kinase have been identified in vertebrates. Expression of these isozymes is tissue-specific and developmentally regulated.
Specifications
Adenylate Kinase 3 | |
Polyclonal | |
AK3 | |
Adenylate kinase isoenzyme 4, mitochondrial; Adenylate kinase 3-like; ATP-AMP transphosphorylase; AK3L1; AK3; AK4 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
50808, 56248 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q9UIJ7, Q9WTP7 | |
AK3 | |
KLH conjugated synthetic peptide between 23-53 amino acids from the N-terminal region of human AK3 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction