Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKAP13 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB10068213
Description
AKAP13 Polyclonal specifically detects AKAP13 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).Specifications
| AKAP13 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| A kinase (PRKA) anchor protein 13, AKAP-LbcAKAP-13, A-kinase anchor protein 13, A-kinase anchoring protein, ARHGEF13, Breast cancer nuclear receptor-binding auxiliary protein, BRXNon-oncogenic Rho GTPase-specific GTP exchange factor, c-lbc, FLJ11952, FLJ43341, Guanine nucleotide exchange factor Lbc, HA-3, HT31, Human thyroid-anchoring protein 31, LBCLBC oncogene, Lymphoid blast crisis oncogeneProtein kinase A-anchoring protein 13, p47, PROTO-LB, PROTO-LBC | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Expected cross-reactivity based on sequence identity (not validated): Guinea pig (100%), Orangutan (100%), Gorilla (100%), Chimpanzee (100%), White-tufted-ear marmoset (100%), Naked mole rat (100%), African elephant (100%), Gray short-tailed opossum (100%), Little brown bat (100%), Northern white-cheeked gibbon (100%), and Small-eared galago (100%). | |
| Store at 4C. Do not freeze. |
| Immunoprecipitation, Western Blot | |
| 1.0 mg/mL | |
| Immunoprecipitation 2-5 ug/mg lysate, Western Blot (Negative) | |
| Q12802 | |
| AKAP13 | |
| The immunogen recognized by this antibody maps to a region between residue 1850 and 1900 of human A kinase (PRKA) anchor protein 13 using the numbering given in entry NP_009131.2 (GeneID 11214). | |
| 100 μL | |
| Breast Cancer | |
| 11214 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction