Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKAP450 Mouse anti-Human, Unlabeled, Clone: 7, BD

Catalog No. BDB611518 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611518 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611518 Supplier BD Biosciences Supplier No. 611518
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Compartmentalization of the type II cAMP-dependent protein kinase (PKA) within the cell is essential for its discrete physiological effects. PKA localization is mediated by interactions between the regulatory (RII) subunit and A-kinase anchoring proteins (AKAPs) which position PKA in close proximity to relevant substrates. Some AKAPs exhibit tissue specific expression, while others are detected ubiquitously. AKAP450 localizes PKA type II to centrosomes. The N-terminal portion of AKAP450 (amino acids 1-1626) is almost identical to the Yotiao protein (∽210 kDa), an AKAP that localizes PKA to NMDA receptors. In addition, AKAP450 contains three major regions (cc1, cc2, cc3) of coiled coil structures, which are characteristic features of centrosomeal proteins. It is expressed at low levels in multiple tissues, including skeletal muscle and liver, but is highly expressed in kidney. PKA is thought to be involved in the maintenance of the interphase microtubule network and to be necessary for stabilization of minus-end microtubules that originate from the centrosome. Thus AKAP450 may serve as a scaffolding protein to localize PKA and other signaling molecules together in cetrosomes.

This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen AKAP450
Applications Western Blot
Classification Monoclonal
Clone 7
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human AKAP450 aa. 17-130
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.