Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKAP79 Mouse anti-Human, Unlabeled, Clone: 22, BD

Catalog No. BDB610314 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610314 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610314 Supplier BD Biosciences Supplier No. 610314

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

The type II cAMP-dependent Protein Kinase (PKA) is compartmentalized within the cell. To maintain this localization of type II PKAs, the regulatory subunit(RII) interacts with specific RII-anchoring proteins. For instance, attachment of type II PKA to the cytoskeleton occurs through the binding of RII to microtubule-associated protein 2 (MAP2). In brain, several proteins have been identified as PKA type II anchoring proteins and form a family named AKAP (A-Kinase Anchor Proteins). AKAP79 is a 79kDa human RII-anchoring protein. AKAP, PKA type II, and calcineurin (PP2B) can form a tertiary complex, suggesting that both PKA and calcineurin are targeted by a common protein to subcellular sites where they regulate the phosphorylation status of key substrates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen AKAP79
Applications Western Blot
Classification Monoclonal
Clone 22
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.