Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKAP82 Mouse, Unlabeled, Clone: 25, BD

Catalog No. BDB611564 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611564 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611564 Supplier BD Biosciences Supplier No. 611564

Mouse Monoclonal Antibody

The cAMP-dependent Protein Kinase (PKA) is compartmentalized within the cell. To maintain the localization of PKA, the regulatory subunits interact with specific anchoring proteins. Several proteins have been identified as PKA anchoring proteins and form a family named AKAP (A-Kinase Anchor Proteins). Fifteen of the AKAP proteins contain a consensus binding motif that allows interaction with the type II regulatory subunit (RII) of the PKA holoenzyme. In addition, three other AKAPs (D-AKAP1, D-AKAP2, and fsc1/AKAP82) can associate with the type I regulatory subunit (RI) of the PKA holoenzyme. AKAP82 was isolated as a component of the mouse sperm fibrous sheath. It is a dual specificity AKAP that contains an RII-binding domain (domain A; amino acids 219 to 232) and an RI-binding domain (domain B; amino acids 335-344). In mouse, pro-AKAP82 is synthesized as a 97 kDa precursor that is transported to the flagellum where proteolytic cleavage of the N-terminal 179 amino acids produces AKAP82. Assembly of AKAP82 into the fibrous sheath surrounding the axoneme of the sperm flagellum is thought to tether PKA close to the axoneme where it can regulate flagellar motility.

TRUSTED_SUSTAINABILITY

Specifications

Antigen AKAP82
Applications Western Blot
Classification Monoclonal
Clone 25
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse AKAP82 aa. 555-675
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.