Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKR1C2 (M03A), Mouse anti-Human, Clone: 3C11, Abnova™

Catalog No. 89021278 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-278 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-278 Supplier Abnova Corporation Supplier No. H00001646M03A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant AKR1C2.

This gene encodes a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. These enzymes catalyze the conversion of aldehydes and ketones to their corresponding alcohols using NADH and/or NADPH as cofactors. The enzymes display overlapping but distinct substrate specificity. This enzyme binds bile acid with high affinity, and shows minimal 3-alpha-hydroxysteroid dehydrogenase activity. This gene shares high sequence identity with three other gene members and is clustered with those three genes at chromosome 10p15-p14. [provided by RefSeq

Sequence: EEPWVDPNSPVLLEDPVLCALAKKHKRTPALIALRYQLQRGVVVLAKSYNEQRIRQNVQVFEFQLTSEEMKAIDGLNRNVRYLTLDIFAGPPNYPVSDEY

Specifications

Antigen AKR1C2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3C11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant AKR1C2.
Formulation ascites with no preservative
Gene AKR1C2
Gene Accession No. BC063574
Gene Alias AKR1C-pseudo/BABP/DD/DD2/DDH2/HAKRD/HBAB/MCDR2
Gene Symbols AKR1C2
Host Species Mouse
Immunogen AKR1C2 (AAH63574, 224 a.a. ∼ 323 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1646
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.