Learn More
aldo-keto reductase family 1, member D1 (delta 4-3-ketosteroid-5-beta-reductase), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human AKR1D1 protein.
Supplier: Abnova Corporation H00006718D01P
Description
The enzyme encoded by this gene is responsible for the catalysis of the 5-beta-reduction of bile acid intermediates and steroid hormones carrying a delta(4)-3-one structure. Deficiency of this enzyme may contribute to hepatic dysfunction. [provided by RefSeq
Sequence: MDLSAASHRIPLSDGNSIPIIGLGTYSEPKSTPKGACATSVKVAIDTGYRHIDGAYIYQNEHEVGEAIREKIAEGKVRREDIFYCGKLWATNHVPEMVRPTLERTLRVLQLDYVDLYIIEVPMAFKPGDEIYPRDENGKWLYHKSNLCATWEAMEACKDAGLVKSLGVSNFNRRQLELILNKPGLKHKPVSNQVECHPYFTQPKLLKFCQQHDIVITAYSPLGTSRNPIWVNVSSPPLLKDALLNSLGKRYNKTAAQIVLRFNIQRGVVVIPKSFNLERIKENFQIFDFSLTEEEMKDIEALNKNVRFVELLMWRDHPEYPFHDEYSpecifications
aldo-keto reductase family 1, member D1 (delta 4-3-ketosteroid-5-beta-reductase) | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human AKR1D1 protein. | |
AKR1D1 | |
3o5bred/SRD5B1 | |
Rabbit | |
Affinity Purified | |
RUO | |
6718 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_005989.2 | |
AKR1D1 | |
AKR1D1 (NP_005980.1, 1 a.a. ∼ 326 a.a) full-length human protein. | |
100 μg | |
Primary | |
Human | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.