Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Akt, PH Domain Clone: SKB1,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ ST10881SET
Description
Specifically detects Akt, PH Domain Clone: SKB1 in Human, Mouse, Rat samples, and it is validated for Flow Cytometry, Immunoprecipitation, Immuno Blotting
Akt (Accession No. NM_005163) is primarily a cytosolic enzyme that acts as a downstream effector of phosphatidylinositol 3-kinase (PI-3K) activation. Akt has been shown to be activated by a variety of stimuli, including hematopoetic cytokines, various chemokines, heat shock, hypoxia,hyperosmolarity, integrins, nerve growth factor (NGF), platelet- derived growth factor (PDGF), and T cell receptor signaling. Suggested to play a role in regulating the balance between cellular mitogenesis and apoptosis. Akt has been shown to be constitutively active in some human cancers.
Specifications
Akt, PH Domain | |
Monoclonal | |
Unconjugated | |
Mouse | |
1 Set | |
Primary | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry, Immunoblot, Immunoblot, Immunoprecipitation | |
SKB1 | |
100μg antibody in 150mM NaCl, 100mM Tris-glycine, 30% glycerol, pH 7.4 and 100μg control lysate. | |
a recombinant protein consisting of amino acids 1-149 of human Akt 1 fused to GST | |
RUO | |
Recognizes the ∽60kDa Akt protein in A431 cells. Useful for the concentration of Akt in samples by immunoprecipitation prior to use in the K-LISA™ Akt Activity Kit. Antibody Target Gene Symbol: AKT1 Target Synonym: AKT, MGC99656, PKB, PKB-ALPHA, PKB/AKT, PRKBA, Protein kinase B, RAC, RAC-ALPHA, Thymoma viral proto-oncogene 1 Entrez Gene Name: v-akt murine thymoma viral oncogene homolog 1 | |
−20°C | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction