Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Akt/PKB Mouse anti-Human, Mouse, Rat, AlexaFluor™488, Clone: SKB1, MilliporeSigma™

Catalog No. 16293MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-293-MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-293-MI Supplier MilliporeSigma Supplier No. 16293
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Akt/PKB Antibody, PH Domain, clone SKB1, Alexa Fluor 488 is an antibody against Akt/PKB for use in flow cytometry, immunocytochemistry, immunofluorescence and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Akt/PKB
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone SKB1
Conjugate Alexa Fluor 488
Gene Accession No. NM_001014431.1; NM_001014432.1; NM_005163.2
Gene Symbols AKT1, RAC, PRKBA, MGC99656, RAC-ALPHA, RAC-PK-alpha, C-AKT, PKB, AKT
Host Species Mouse
Immunogen GST-fusion protein corresponding to residues 1-149 of human Akt1. Clone SKB1.
Purification Method Protein G purfied
Quantity 50 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 4°C for one year
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.