Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Akt/PKB Mouse anti-Human, Mouse, Rat, Clone: SKB1, MilliporeSigma™

Catalog No. 16318MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-318-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-318-MI Supplier MilliporeSigma Supplier No. 16318
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Akt/PKB using this Anti-Akt/PKB Antibody, PH Domain, clone SKB1, Magnetic Bead Conjugate validated for use in immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Akt/PKB
Applications Immunoprecipitation
Classification Monoclonal
Clone SKB1
Conjugate Unconjugated
Formulation 20% slurry; 1X PBS, 0.05% sodium azide.
Sufficient for 10 IPs.
Host Species Mouse
Immunogen GST-fusion protein corresponding to residues 1-149 of human Akt1
Purification Method Protein G Chromatography
Quantity 200 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.