Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Akt1/PKBα Rabbit anti-Human, Mouse, Rat, Clone: AW24, MilliporeSigma™

Catalog No. 04796MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-796-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-796-MI Supplier MilliporeSigma Supplier No. 04796
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Anti-Akt1 Antibody, clone AW24 detects level of Akt1/PKBα and has been published and validated for use in western blotting and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Akt1/PKBα
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone AW24
Conjugate Unconjugated
Formulation Cultured supernantant containing 0.05% sodium azide.
Gene Accession No. NP_150233.1
Host Species Rabbit
Immunogen Sythetic peptide (C-RPHFPQFSYSASGTA) corresponding to the C-terminal (residues 466-480) of rat Akt1/PKBα.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) AKT, C-AKT, EC 2.7.11.1, MGC99656, PKB, PKB-ALPHA, PRKBA, RAC, RAC-ALPHA, RAC-PK-alpha
Target Species Human, Mouse, Rat
Content And Storage −20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.