Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Akt1/PKBα Rabbit anti-Human, Rat, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 07416MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-416-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-416-MI Supplier MilliporeSigma Supplier No. 07416
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Akt1/PKBα Antibody is validated for use in immunoprecipitation, western blotting for the detection of Akt1/PKBα.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Akt1/PKBα
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%.
Gene Symbols AKT1; RAC; PRKBA; RAC-ALPHA; PKB; MGC99656; RAC-PK-alpha; C-AKT; AKT
Host Species Rabbit
Immunogen 15 residue synthetic peptide (C-RPHFPQFS YSASGTA) corresponding to the C-terminal (residues 466-480) of rat Akt1, with an N-terminal cysteine added for conjugation purposes. The Akt1 sequence is identical to mouse Akt1 (15/15 amino acids) and strongly co
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_001014431
Target Species Human, Mouse, Rat
Content And Storage −20°C for up to 12 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.