Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Akt2/PKBΒ Rabbit anti-Human, Rat, Clone: AW114, MilliporeSigma™

Catalog No. 05771MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-771-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-771-MI Supplier MilliporeSigma Supplier No. 05771

Rabbit Monoclonal Antibody

Detect Akt2/PKBβ using this Anti-Akt2/PKBβ Antibody, clone AW114 validated for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Akt2/PKBβ
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone AW114
Conjugate Unconjugated
Formulation 0.1M Tris-Glycine, 0.15M NaCl, 0.05% Sodium Azide, pH 7.4.
Gene Accession No. NM_001626
Host Species Rabbit
Immunogen Peptide (C-RYDSLGSLELDQRTH) corresponding to amino acids 455-469 of human Akt2/PKBb
Purification Method Protein A purfied
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) AKT2, RAC-BETA, PRKBB, PKBBETA, RAC-PK-beta
Target Species Human, Rat
Content And Storage 2-8°C for 12 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.