Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

v-akt murine thymoma viral oncogene homolog 3 (protein kinase B, gamma), Mouse, Clone: 6E10, Abnova™

Catalog No. 89014247 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-247 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-247 Supplier Abnova Corporation Supplier No. H00010000M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant AKT3.

The protein encoded by this gene is a member of the AKT, also called PKB, serine/threonine protein kinase family. AKT kinases are known to be regulators of cell signaling in response to insulin and growth factors. They are involved in a wide variety of biological processes including cell proliferation, differentiation, apoptosis, tumorigenesis, as well as glycogen synthesis and glucose uptake. This kinase has been shown to be stimulated by platelet-derived growth factor (PDGF), insulin, and insulin-like growth factor 1 (IGF1). Alternatively splice transcript variants encoding distinct isoforms have been described. [provided by RefSeq

Sequence: EAIQAVADRLQRQEEERMNCSPTSQIDNIGEEEMDASTTHHKRKTMNDFDYLKLLGKGTFGKVILVREKASGKYYAMKILKKEVIIAKDE

Specifications

Antigen v-akt murine thymoma viral oncogene homolog 3 (protein kinase B, gamma)
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 6E10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant AKT3.
Formulation PBS with no preservative; pH 7.4
Gene AKT3
Gene Accession No. AF124141
Gene Alias DKFZp434N0250/PKB-GAMMA/PKBG/PRKBG/RAC-PK-gamma/RAC-gamma/STK-2
Gene Symbols AKT3
Host Species Mouse
Immunogen AKT3 (AAD29089, 100 a.a. ∼ 189 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Kinases
Primary or Secondary Primary
Gene ID (Entrez) 10000
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.