Learn More
ALAS2, Mouse, Clone: 4D8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ALAS2.
Supplier: Abnova Corporation H00000212M02
Description
The product of this gene specifies an erythroid-specific mitochondrially located enzyme. The encoded protein catalyzes the first step in the heme biosynthetic pathway. Defects in this gene cause X-linked pyridoxine-responsive sideroblastic anemia. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq
Sequence: MVTAAMLLQCCPVLARGPTSLLGKVVKTHQFLFGIGRCPILATQGPNCSQIHLKATKAGGDSPSWAKGHCPFMLSELQDGKSKIVQKAAPEVQEDVKAFKSpecifications
ALAS2 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_000032 | |
ALAS2 | |
ALAS2 (NP_000023, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
4D8 | |
Mouse monoclonal antibody raised against a partial recombinant ALAS2. | |
ALAS2 | |
ALAS-E/ALASE/ANH1/ASB/FLJ93603/XLSA | |
Mouse | |
Affinity chromatography | |
RUO | |
212 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.