Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Albumin Serum, APC, Clone: 188835, R&D Systems™

Catalog No. IC1455A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC1455A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC1455A Supplier R&D Systems Supplier No. IC1455A
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Host Species: Mouse
Clone: 188835
Isotype: IgG2a
Species Reactivity: Hu, Ms
Immunogen: Human Serum Albumin

Applications:

Flow Cytometry (Intracellular Staining)

Specifications

Antigen Albumin
Applications Flow Cytometry
Classification Monoclonal
Clone 188835
Conjugate APC
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Gene Alias ALB, albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
Gene Symbols ALB
Host Species Mouse
Immunogen Human Serum Albumin
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) 213
Test Specificity Detects human Serum Albumin in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.