Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ALCAM/CD166 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB6561G
This item is not returnable.
View return policy
Description
ALCAM/CD166 Monoclonal specifically detects ALCAM/CD166 in Human samples. It is validated for Flow Cytometry.Specifications
ALCAM/CD166 | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
activated leucocyte cell adhesion molecule, activated leukocyte cell adhesion moleculeFLJ38514, CD166, CD166 antigen, MEMDMGC71733 | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
214 | |
Human | |
Purified |
Flow Cytometry | |
105902 | |
Flow Cytometry 5 uL/10^6 cells | |
Q13740 | |
ALCAM | |
Mouse myeloma cell line NS0-derived recombinant human ALCAM/CD166 Trp28-Ala526 Accession # Q13740 | |
100 Tests | |
Primary | |
Detects human ALCAM in Western blots. Shows approximately 50% cross-reactivity with recombinant mouse OCAM and no cross-reactivity with recombinant human (rh) BCAM, rhEpCAM, rhMCAM, or rhNCAM-L1. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction