Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDH1B1, Mouse, Clone: 2C7, Abnova™

Catalog No. 89016112 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-112 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-112 Supplier Abnova Corporation Supplier No. H00000219M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant ALDH1B1.

This protein belongs to the aldehyde dehydrogenases family of proteins. Aldehyde dehydrogenase is the second enzyme of the major oxidative pathway of alcohol metabolism. This gene does not contain introns in the coding sequence. The variation of this locus may affect the development of alcohol-related problems. [provided by RefSeq

Sequence: MLRFLAPRLLSLQGRTARYSSAAALPSPILNPDIPYNQLFINNEWQDAVSKKTFPTVNPTTGEVIGHVAEGDRADVDRAVKAAREAFRLGSPWRRMDASERGRLLNLLADLVERDRVYLASLETLDNGKPFQESYALDLDEVIKVYRYFAGWADKWHGKTIPMDGQHFCFTRHEPVGVCGQIIPWNFPLVMQGWKLAPALATGNTVVMKVAEQTPLSALYLASLIKEAGFPPGVVNIITGYGPTAGAAIAQHMDVDKVAFTGSTEVGHLIQKAAGDSNLKRVTLELGGKSPSIVLADADMEHAVEQCHEALFFNMGQCCCAGSRTFVEESIYNEFLERTVEKAKQRKVGNPFELDTQQGPQVDKEQFERVLGYIQLGQKEGAKLLCGGERFGERGFFIKPTVFGGVQDDMRIAKEEIFGPVQPLFKFKKIEEVVERANNTRYGLAAAVFTRDLDKAMYFTQALQAGTVWVNTYNIVTCHTPFGGFKESGNGRELGEDGLKAYTEVKTVTIKVPQKNS

Specifications

Antigen ALDH1B1
Applications ELISA
Classification Monoclonal
Clone 2C7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant ALDH1B1.
Formulation PBS with no preservative; pH 7.4
Gene ALDH1B1
Gene Accession No. BC001619
Gene Alias ALDH5/ALDHX/MGC2230
Gene Symbols ALDH1B1
Host Species Mouse
Immunogen ALDH1B1 (AAH01619, 1 a.a. ∼ 517 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.