Learn More
aldehyde dehydrogenase 3 family, memberA1, Mouse, Clone: 2G4, Abnova™
Shop All Abnova Corporation ProductsDescription
Aldehyde dehydrogenases oxidize various aldehydes to the corresponding acids. They are involved in the detoxification of alcohol-derived acetaldehyde and in the metabolism of corticosteroids, biogenic amines, neurotransmitters, and lipid peroxidation. The enzyme encoded by this gene forms a cytoplasmic homodimer that preferentially oxidizes aromatic and medium-chain (6 carbons or more) saturated and unsaturated aldehyde substrates. It is thought to promote resistance to UV and 4-hydroxy-2-nonenal-induced oxidative damage in the cornea. The gene is located within the Smith-Magenis syndrome region on chromosome 17. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq
Specifications
Specifications
| Antigen | aldehyde dehydrogenase 3 family, memberA1 |
| Applications | ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 2G4 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a full length recombinant ALDH3A1. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ALDH3A1 |
| Gene Accession No. | BC004370 |
| Gene Alias | ALDH3/ALDHIII/MGC10406 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.