Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

aldehyde dehydrogenase 3 family, memberA1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016706 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-706 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-706 Supplier Abnova Corporation Supplier No. H00000218B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human ALDH3A1 protein.

Aldehyde dehydrogenases oxidize various aldehydes to the corresponding acids. They are involved in the detoxification of alcohol-derived acetaldehyde and in the metabolism of corticosteroids, biogenic amines, neurotransmitters, and lipid peroxidation. The enzyme encoded by this gene forms a cytoplasmic homodimer that preferentially oxidizes aromatic and medium-chain (6 carbons or more) saturated and unsaturated aldehyde substrates. It is thought to promote resistance to UV and 4-hydroxy-2-nonenal-induced oxidative damage in the cornea. The gene is located within the Smith-Magenis syndrome region on chromosome 17. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq

Sequence: MSKISEAVKRARAAFSSGRTRPLQFRIQQLEALQRLIQEQEQELVGALAADLHKNEWNAYYEEVVYVLEEIEYMIQKLPEWAADEPVEKTPQTQQDELYIHSEPLGVVLVIGTWNYPFNLTIQPMVGAIAAGNAVVLKPSELSENMASLLATIIPQYLDKDLYPVINGGVPETTELLKERFDHILYTGSTGVGKIIMTAAAKHLTPVTLELGGKSPCYVDKNCDLDVACRRIAWGKFMNSGQTCVAPDYILCDPSIQNQIVEKLKKSLKEFYGEDAKKSRDYGRIISARHFQRVMGLIEGQKVAYGGTGDAATRYIAPTILTDVDPQSPVMQEEIFGPVLPIVCVRSLEEAIQFINQREKPLALYMFSSNDKVIKKMIAETSSGGVAANDVIVHITLHSLPFGGVGNSGMGSYHGKKSFETFSHRRSCLVRPLMNDEGLKVRYPPSPAKMTQH

Specifications

Antigen aldehyde dehydrogenase 3 family, memberA1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human ALDH3A1 protein.
Formulation PBS with no preservative; pH 7.4
Gene ALDH3A1
Gene Accession No. BC008892.2
Gene Alias ALDH3/ALDHIII/MGC10406
Gene Symbols ALDH3A1
Host Species Mouse
Immunogen ALDH3A1 (AAH08892.1, 1 a.a. ∼ 453 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 218
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.