Learn More
ALDH3B1, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human ALDH3B1 peptide using ARM Technology.
Supplier: Abnova Corporation H00000221K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
The aldehyde dehydrogenases are a family of isozymes that may play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. This particular gene spans about 20kb of genomic DNA and is composed of 9 coding exons. The gene is highly expressed in kidney and lung. The functional significance of this gene as well as the cellular localization of its product are presently unknown. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
ALDH3B1 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
ALDH3B1 | |
ALDH3B1 | |
A synthetic peptide of human ALDH3B1 | |
RUO | |
221 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
aldehyde dehydrogenase 3 family, member B1 | |
ALDH3B1 | |
ALDH4, ALDH7, FLJ26433 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.