Learn More
aldehyde dehydrogenase 6 family, member A1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human ALDH6A1 protein.
Supplier: Abnova Corporation H00004329B01
Description
This protein belongs to the aldehyde dehydrogenases family of proteins. This enzyme plays a role in the valine and pyrimidine catabolic pathways. The product of this gene, a mitochondrial methylmalonate semialdehyde dehydrogenase, catalyzes the irreversible oxidative decarboxylation of malonate and methylmalonate semialdehydes to acetyl- and propionyl-CoA. Methylmalonate semialdehyde dehydrogenase deficiency is characterized by elevated beta-alanine, 3-hydroxypropionic acid, and both isomers of 3-amino and 3-hydroxyisobutyric acids in urine organic acids. [provided by RefSeq
Sequence: MAALLAAAAVRARILQVSSKVKSSPTWYSASSFSSSVPTVKLFIGGKFVESKSDKWIDIHNPATNEVIGRVPQATKAEMDAAIASCKRAFPAWADTSVLSRQQVLLRYQQLIKENLKEIAKLITLEQGKTLADAEGDVFRGLQVVEHACSVTSLMMGETMPSITKDMDLYSYRLPLGVCAGIAPFNFPAMIPLWMFPMAMVCGNTFLMKPSERVPGATMLLAKLLQDSGAPDGTLNIIHGQHEAVNFICDHPDIKAISFVGSNKAGEYIFERGSRHGKRVQANMGAKNHGVVMPDANKENTLNQLVGAAFGAAGQRCMALSTAVLVGEAKKWLPELVEHAKNLRVNAGDQPGADLGPLITPQAKERVCNLIDSGTKEGASILLDGRKIKVKGYENGNFVGPTIISNVKPNMTCYKEEIFGPVLVVLETETLDEAIQIVNNNPYGNGTAIFTTNGATARKYAHLVDVGQVGVNVPIPVPLPMFSFTGSRSSFRGDTNFYGKQGIQFYTQLKTITSQWKEEDATLSSPAVVMPTMGRSpecifications
aldehyde dehydrogenase 6 family, member A1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human ALDH6A1 protein. | |
ALDH6A1 | |
MGC40271/MMSADHA/MMSDH | |
Mouse | |
50 μL | |
Yes | |
4329 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
No additive | |
NM_005589.2 | |
ALDH6A1 | |
ALDH6A1 (NP_005580.1, 1 a.a. ∼ 535 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.