Learn More
aldolase A, fructose-bisphosphate, Mouse, Clone: 3D9-6F3, Abnova™
Shop All Abnova Corporation ProductsDescription
This gene product, Aldolase A (fructose-bisphosphate aldolase) is a glycolytic enzyme that catalyzes the reversible conversion of fructose-1,6-bisphosphate to glyceraldehyde 3-phosphate and dihydroxyacetone phosphate. Three aldolase isozymes (A, B, and C), encoded by three different genes, are differentially expressed during development. Aldolase A is found in the developing embryo and is produced in even greater amounts in adult muscle. Aldolase A expression is repressed in adult liver, kidney and intestine and similar to aldolase C levels in brain and other nervous tissue. Aldolase A deficiency has been associated with myopathy and hemolytic anemia. Alternative splicing of this gene results in multiple transcript variants which encode the same protein. [provided by RefSeq
Specifications
Specifications
| Antigen | aldolase A, fructose-bisphosphate |
| Applications | ELISA, Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | 3D9-6F3 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a full length recombinant ALDOA. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ALDOA |
| Gene Accession No. | BC010660 |
| Gene Alias | ALDA/MGC10942/MGC17716/MGC17767 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.