Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Anti- Alemtuzumab (Anti-Idiotype) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $456.00
Specifications
| Antigen | Alemtuzumab |
|---|---|
| Clone | 2499A |
| Dilution | Flow Cytometry 0.25 μg/106 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10246SP
![]() |
R&D Systems
MAB10246SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10246100
![]() |
R&D Systems
MAB10246100 |
100 μg |
Each for $456.00
|
|
|||||
Description
Alemtuzumab Monoclonal specifically detects Alemtuzumab in Multi-Species samples. It is validated for Flow Cytometry.Specifications
| Alemtuzumab | |
| Flow Cytometry 0.25 μg/106 cells | |
| Monoclonal | |
| Supernatant | |
| Multi-species | |
| Full-length Alemtuzumab | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects Alemtuzumab in direct ELISAs and Western blots. |
| 2499A | |
| Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title