Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

asparagine-linked glycosylation 1, beta-1,4-mannosyltransferase homolog (S. cerevisiae), Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004679 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-679 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-679 Supplier Abnova Corporation Supplier No. H00056052A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant ALG1.

The enzyme encoded by this gene catalyzes the first mannosylation step in the biosynthesis of lipid-linked oligosaccharides. This gene is mutated in congenital disorder of glycosylation type Ik. [provided by RefSeq

Sequence: IDWHNYGYSIMGLVHGPNHPLVLLAKWYEKFFGRLSHLNLCVTNAMREDLADNWHIRAVTVYDKPASFFKETPLDLQHRLFMKLGSMHSPFRARSEPEDP

Specifications

Antigen asparagine-linked glycosylation 1, beta-1,4-mannosyltransferase homolog (S. cerevisiae)
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant ALG1.
Formulation 50% glycerol
Gene ALG1
Gene Accession No. NM_019109
Gene Alias HMAT1/HMT-1/HMT1
Gene Symbols ALG1
Host Species Mouse
Immunogen ALG1 (NP_061982, 154 a.a. ∼ 253 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 56052
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.