Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alkaline Phosphatase Mouse anti-Human, Alexa Fluor 488, Clone: B4-78, BD

Catalog No. BDB561495 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561495 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561495 Supplier BD Biosciences Supplier No. 561495

Mouse Monoclonal Antibody

The B4-78 monoclonal antibody reacts with the tissue-nonspecific isozyme of alkaline phosphatase. Alkaline phosphatases are membrane-bound glycoproteins. Four isozymes of alkaline phosphatase exist in humans: placental, placental-like, intestinal, and liver/bone/kidney. Liver/bone/kidney alkaline phosphatase is also known as tissue-nonspecific alkaline phosphatase (TNAP). Human embryonic stem cells and embryonic carcinoma cells express high levels of tissue-nonspecific alkaline phosphatase that decrease upon differentiation. Genetic and biochemical studies suggest that TNAP plays a role in skeletal mineralization.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Alkaline Phosphatase
Applications Flow Cytometry
Classification Monoclonal
Clone B4-78
Conjugate Alexa Fluor 488
Description Bone/Kidney/Liver Alkaline Phosphatase, TNAP, TNSALP, AP-TNAP, ALPL
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Bone Alkaline Phosphatase
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.