Learn More
alkB, alkylation repair homolog 1 (E. coli), Mouse, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
This gene encodes a homolog to the E. coli alkB gene product. The E. coli alkB protein is part of the adaptive response mechanism of DNA alkylation damage repair. It is involved in damage reversal by oxidative demethylation of 1-methyladenine and 3-methylcytosine. [provided by RefSeq
Specifications
Specifications
| Antigen | alkB, alkylation repair homolog 1 (E. coli) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a full-length human ALKBH protein. |
| Formulation | No additive |
| Gene | ALKBH1 |
| Gene Accession No. | BC025787.1 |
| Gene Alias | ABH/ABH1/ALKBH/alkB/hABH |
| Gene Symbols | ALKBH1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.